missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-1-amine, tech. 90%

Catalog No. AAL2015703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2015703 1 g
AAL2015706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2015703 Supplier Thermo Scientific Chemicals Supplier No. L2015703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2524-49-4 | C4H9N | 71.12 g/mol

3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.

Solubility
Miscible with water.

Notes
Incompatible with strong acids, strong oxidizing agents and strong bases.

Chemical Identifiers

CAS 2524-49-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD03425859
InChI Key ASVKKRLMJCWVQF-UHFFFAOYSA-N
Synonym 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
PubChem CID 443732
ChEBI CHEBI:31108
IUPAC Name but-3-en-1-amine
SMILES NCCC=C

Specifications

Density 0.777
Boiling Point 74°C to 77°C
Flash Point −5°C (23°F)
Odor Amine-like
Refractive Index 1.424
Quantity 1 g
UN Number UN2733
Beilstein 1735803
Sensitivity Air Sensitive
Solubility Information Miscible with water.
Formula Weight 71.12
Percent Purity 97%
Chemical Name or Material 3-Buten-1-amine
Show More Show Less
Hazard Category H225-H314-H335
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only