missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene 97.0+%, TCI America™

Catalog No. B106725G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B106725G 25 g
B1067250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B106725G Supplier TCI America Supplier No. B106725G
Only null left

Chemical Identifiers

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1

Specifications

Boiling Point 158°C
Quantity 25 g
UN Number 1992
Formula Weight 163.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromothiophene