missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylamine hydrobromide, 98%, Thermo Scientific™

Catalog No. AC107381000
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC107381000 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC107381000 Supplier Thermo Scientific Chemicals Supplier No. 107381000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 5003-71-4
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H8BrN·HBr
Linear Formula Br(CH2)3NH2·HBr
Quantity 100g
MDL Number MFCD00012912
Beilstein 04, 149
Synonym 3-bromopropylamine hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-aminopropyl bromide hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-bromopropylammonium bromide, 3-bromopropanamine hydrobromide, 1-amino-3-bromopropane hydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-brominepropylaminehydrobromide, 3-bromopropylamine, hydrobromide
Solubility Information Solubility in water: soluble
InChI Key PQIYSSSTRHVOBW-UHFFFAOYSA-N
SMILES C(CN)CBr.Br
IUPAC Name 3-bromopropan-1-amine;hydrobromide
Molecular Weight (g/mol) 218.93
PubChem CID 78701
Formula Weight 218.93
Percent Purity 98%
Chemical Name or Material 3-Bromopropylamine hydrobromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 225-675-0
RTECSNumber UH9925000
TSCA TSCA

RUO – Research Use Only