missing translation for 'onlineSavingsMsg'
Learn More

(3-Bromopropoxy)(tert-butyl)dimethylsilane 98.0+%, TCI America™

Catalog No. B53465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B53465G 5 g
B534625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B53465G Supplier TCI America Supplier No. B53465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89031-84-5
Molecular Formula C9H21BrOSi
Molecular Weight (g/mol) 253.255
MDL Number MFCD00216589
InChI Key QGMROEZDWJTIDW-UHFFFAOYSA-N
Synonym 3-bromopropoxy-tert-butyldimethylsilane, 3-bromopropoxy tert-butyl dimethylsilane, 3-bromopropoxy-t-butyldimethylsilane, 3-bromopropoxy-tert-butyl-dimethyl-silane, silane, 3-bromopropoxy 1,1-dimethylethyl dimethyl, acmc-209unq, t-butyl 3-bromopropoxy dimethylsilane, bromopropoxy-tert-butyldimethyl silane
PubChem CID 607486
IUPAC Name 3-bromopropoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCCCBr

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 253.26
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (3-Bromopropoxy)(tert-butyl)dimethylsilane
TSCA No
Recommended Storage Refrigerator