missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenylhydrazine hydrochloride, 98%

Catalog No. AC167350250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC167350250 25 g Glass bottle
AC167350050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC167350250 Supplier Thermo Scientific Chemicals Supplier No. 167350250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 27246-81-7 | C6H8BrClN2 | 223.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27246-81-7
Molecular Formula C6H8BrClN2
Molecular Weight (g/mol) 223.50
MDL Number MFCD00012933
InChI Key RPYIPFXHIKXRKS-UHFFFAOYSA-N
Synonym 3-bromophenylhydrazine hydrochloride, 3-bromophenyl hydrazine hydrochloride, 1-3-bromophenyl hydrazine hydrochloride, m-bromophenyl hydrazine hydrochloride, hydrazine, m-bromophenyl-, monohydrochloride, 3-bromophenylhydrazinehydrochloride, 3-bromophenyl hydrazine hcl, hydrazine, 3-bromophenyl-, monohydrochloride, pubchem23212, acmc-1clhx
PubChem CID 33757
IUPAC Name (3-bromophenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].NNC1=CC=CC(Br)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 225°C to 231°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula BrC6H4NHNH2·HCl
Quantity 25 g
Beilstein 15, 433
Formula Weight 223.51
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Bromophenylhydrazine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 248-362-

RUO – Research Use Only