missing translation for 'onlineSavingsMsg'
Learn More

3-Bromophenetole 98.0+%, TCI America™

Catalog No. B20625G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20625G 5 g
B206225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20625G Supplier TCI America Supplier No. B20625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2655-84-7
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00274287
InChI Key LQBMPJSTUHWGDE-UHFFFAOYSA-N
Synonym 3-bromophenetole, benzene, 1-bromo-3-ethoxy, m-bromophenetole, 3-bromophenyl ethyl ether, 1-bromo-3-ethoxy-benzene, 3-bromoethoxybenzene, 3-bromphenyl ethyl ether, phenetole, m-bromo, m-bromphenetol, 3-ethoxybromobenzene
PubChem CID 75867
IUPAC Name 1-bromo-3-ethoxybenzene
SMILES CCOC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 231°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromophenetole
EINECSNumber (3)-0845
TSCA No