missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™

Catalog No. CC25410DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC25410DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC25410DA Supplier Thermo Scientific Chemicals Supplier No. CC25410DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4926-47-0
Molecular Formula C7H5BrN2
Molecular Weight (g/mol) 197.035
InChI Key APYSHMNJHJRIDR-UHFFFAOYSA-N
PubChem CID 327951
IUPAC Name 3-bromoimidazo[1,2-a]pyridine
SMILES C1=CC2=NC=C(N2C=C1)Br

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Synonym 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo
Formula Weight 197.03
Percent Purity 97%
Chemical Name or Material 3-Bromoimidazo[1, 2-a]pyridine