missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluorobenzene 98.0+%, TCI America™

Catalog No. B088225G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B088225G 25 g
B0882250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B088225G Supplier TCI America Supplier No. B088225G

Chemical Identifiers

CAS 1073-06-9
Molecular Formula C6H4BrF
Molecular Weight (g/mol) 175
MDL Number MFCD00000326
InChI Key QDFKKJYEIFBEFC-UHFFFAOYSA-N
Synonym 3-bromofluorobenzene, m-bromofluorobenzene, m-fluorobromobenzene, benzene, 1-bromo-3-fluoro, 3-bromfluorbenzen, 3-fluorobromobenzene, m-fluorophenyl bromide, 1-fluoro-3-bromobenzene, 3-fluoro bromobenzene, benzene, 1-fluoro-3-bromo
PubChem CID 14082
IUPAC Name 1-bromo-3-fluorobenzene
SMILES C1=CC(=CC(=C1)Br)F

Specifications

Color Yellow
Boiling Point 147°C
Quantity 25 g
UN Number 1993
Formula Weight 175.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromofluorobenzene
EINECSNumber (3)-4286
RTECSNumber DA1225000
TSCA Yes