missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzyl Bromide 98.0+%, TCI America™

Catalog No. B15705G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15705G 5 g
B157025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15705G Supplier TCI America Supplier No. B15705G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 823-78-9
Molecular Formula C7H6Br2
Molecular Weight (g/mol) 249.933
MDL Number MFCD00000176
InChI Key ZPCJPJQUVRIILS-UHFFFAOYSA-N
Synonym 3-bromobenzyl bromide, 1-bromo-3-bromomethyl benzene, m-bromobenzyl bromide, 3-bromobenzylbromide, benzene, 1-bromo-3-bromomethyl, alpha,3-dibromotoluene, alpha,m-dibromotoluene, 3-bromo benzyl bromide, 1-bromo-3-bromomethyl-benzene, alpha-3-dibromotoluene
PubChem CID 69979
IUPAC Name 1-bromo-3-(bromomethyl)benzene
SMILES C1=CC(=CC(=C1)Br)CBr

Specifications

Melting Point 42°C
Boiling Point 135°C
Quantity 5 g
UN Number 3261
Formula Weight 249.93
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzyl Bromide
TSCA Yes
Recommended Storage Refrigerator