missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzotrifluoride, 98+%

CAS: 401-78-5 | C7H4BrF3 | 225.008 g/mol

$63.03 - $729.07

Chemical Identifiers

CAS 401-78-5
Molecular Formula C7H4BrF3
Molecular Weight (g/mol) 225.008
MDL Number MFCD00000380
InChI Key NNMBNYHMJRJUBC-UHFFFAOYSA-N
Synonym 3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
PubChem CID 9817
IUPAC Name 1-bromo-3-(trifluoromethyl)benzene
SMILES C1=CC(=CC(=C1)Br)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1472414
View Documents
Thermo Scientific Chemicals
A1472414
25 g
Each for $63.03
Only null left
 
AAA1472422
View Documents
Thermo Scientific Chemicals
A1472422
100 g
Each for $192.42
Only null left
 
AAA1472436
View Documents
Thermo Scientific Chemicals
A1472436
500 g
Each for $729.07
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

401-78-5
225.008
NNMBNYHMJRJUBC-UHFFFAOYSA-N
9817
C1=CC(=CC(=C1)Br)C(F)(F)F
C7H4BrF3
MFCD00000380
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
1-bromo-3-(trifluoromethyl)benzene

Specifications

401-78-5
151°C to 152°C
Aromatic
1.473
25 g
1449557
NNMBNYHMJRJUBC-UHFFFAOYSA-N
1-bromo-3-(trifluoromethyl)benzene
9817
≥98%
1.613
59°C (138°F)
C7H4BrF3
MFCD00000380
UN1993
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
C1=CC(=CC(=C1)Br)C(F)(F)F
225.008
225.01
3-Bromobenzotrifluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 206-932-6

RTECSNumber : XS7970000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only