missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzotrifluoride, 98+%

CAS: 401-78-5 | C7H4BrF3 | 225.008 g/mol

Supplier:  Thermo Scientific Chemicals A1472422

 View more versions of this product

Catalog No. AAA1472422


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Bromobenzotrifluoride
1.613
59°C (138°F)
C7H4BrF3
MFCD00000380
UN1993
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
C1=CC(=CC(=C1)Br)C(F)(F)F
225.008
225.01
401-78-5
151°C to 152°C
Aromatic
1.473
100 g
1449557
NNMBNYHMJRJUBC-UHFFFAOYSA-N
1-bromo-3-(trifluoromethyl)benzene
9817
≥98%

Chemical Identifiers

401-78-5
225.008
NNMBNYHMJRJUBC-UHFFFAOYSA-N
9817
C1=CC(=CC(=C1)Br)C(F)(F)F
C7H4BrF3
MFCD00000380
3-bromobenzotrifluoride, 1-bromo-3-trifluoromethyl benzene, m-bromobenzotrifluoride, 3-trifluoromethyl bromobenzene, benzene, 1-bromo-3-trifluoromethyl, 3-brombenzotrifluorid, 3-bromotrifluoromethylbenzene, 3-bromobenzyltrifluoride, 3-trifluoromethyl phenyl bromide, m-trifluoromethyl bromobenzene
1-bromo-3-(trifluoromethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 206-932-6

RTECSNumber : XS7970000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only