missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromobenzonitrile, 99%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
Spécifications
| Nom ou substance chimique | 3-Bromobenzonitrile |
| Nom Remarque | 99% |
| CAS % minimum | 98.5 |
| CAS % maximal | 100.0 |
| Point d’ébullition | 225°C |
| Point d’éclair | >110°C |
| Spectre infrarouge | Authentic |
| Fourchette de pourcentages d’essai | 98.5% min. (GC) |
| Emballage | Glass bottle |
| Formule linéaire | BrC6H4CN |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement: Harmful in contact with skin. Harmful to aquatic life with long lasting effects. Causes serious eye irritation. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
| EINECSNumber | 230-127-9 |
| RTECSNumber | DI2459500 |
| TSCA | TSCA |
RUO – Research Use Only