missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzamide 98.0+%, TCI America™

Catalog No. B51785G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51785G 5 g
B517825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51785G Supplier TCI America Supplier No. B51785G

Chemical Identifiers

CAS 22726-00-7
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00017127
InChI Key ODJFDWIECLJWSR-UHFFFAOYSA-N
Synonym m-bromobenzamide, benzamide, 3-bromo, 3-bromo-benzamide, benzamide, m-bromo, 3-carbamoyl bromobenzene, 3-bromanylbenzamide, pubchem3734, 3-bromophenylcarboxamide, acmc-209fyc, 3-bromobenzoic acid amide
PubChem CID 89807
IUPAC Name 3-bromobenzamide
SMILES C1=CC(=CC(=C1)Br)C(=O)N

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 200.04
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromobenzamide