missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromoacetophenone 98.0+%, TCI America™

Catalog No. B05365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B05365G 5 g
B053625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B05365G Supplier TCI America Supplier No. B05365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2142-63-4
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD00000083
InChI Key JYAQYXOVOHJRCS-UHFFFAOYSA-N
Synonym 3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo
PubChem CID 16502
IUPAC Name 1-(3-bromophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)Br

Specifications

Color Yellow
Boiling Point 136°C
Quantity 5 g
Formula Weight 199.05
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Bromoacetophenone