missing translation for 'onlineSavingsMsg'
Learn More

3'-Bromoacetophenone, 98+%

Catalog No. AAA1245414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1245414 25 g
AAA1245422 100 g
AAA1245436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1245414 Supplier Thermo Scientific Chemicals Supplier No. A1245414

CAS: 2142-63-4 | C8H7BrO | 199.047 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2142-63-4
Molecular Formula C8H7BrO
Molecular Weight (g/mol) 199.047
MDL Number MFCD00000083
InChI Key JYAQYXOVOHJRCS-UHFFFAOYSA-N
Synonym 3'-bromoacetophenone, m-bromoacetophenone, 1-3-bromophenyl ethanone, 1-3-bromophenyl ethan-1-one, ethanone, 1-3-bromophenyl, 1-acetyl-3-bromobenzene, 3-bromoacetophenone, 1-3-bromo-phenyl-ethanone, 3-bromo acetophenone, acetophenone, 3'-bromo
PubChem CID 16502
IUPAC Name 1-(3-bromophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)Br

Specifications

Melting Point 7°C to 8°C
Density 1.498
Boiling Point 79°C to 81°C (2 mmHg)
Flash Point 114°C (237°F)
Refractive Index 1.575
Quantity 25 g
Beilstein 507599
Formula Weight 199.05
Percent Purity ≥98%
Chemical Name or Material 3'-Bromoacetophenone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-396-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only