missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-6-methoxy-2-methylpyridine 98.0+%, TCI America™
Spécifications
Spécifications
| Color | Yellow |
| Boiling Point | 86°C |
| Quantity | 1 g |
| Formula Weight | 202.05 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 3-Bromo-6-methoxy-2-methylpyridine |
Sécurité et manipulation
| TSCA | No |