missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-hydroxy-2-methylpyridine 98.0+%, TCI America™

Numéro de catalogue. B27961G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B27961G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. B27961G Fournisseur TCI America Code fournisseur B27961G
Il en reste null

Identifiants chimiques

CAS 54923-31-8
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD03427653
InChI Key UJHCRBDEJPQFIA-UHFFFAOYSA-N
Synonym 3-bromo-6-hydroxy-2-methylpyridine, 5-bromo-6-methylpyridin-2-ol, 3-bromo-6-hydroxy-2-picoline, 5-bromo-2-hydroxy-6-picoline, 5-bromo-6-methyl-2 1h-pyridinone, 2 1h-pyridinone, 5-bromo-6-methyl, 5-bromo-2-hydroxy-6-methylpyridine, 5-bromo-6-methyl-2-pyridinol, 5-bromo-6-methylpyridin-2 1h-one, acmc-20aiyp
PubChem CID 2734417
IUPAC Name 5-bromo-6-methyl-1,2-dihydropyridin-2-one
SMILES CC1=C(Br)C=CC(=O)N1

Spécifications

Melting Point 202°C
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-6-hydroxy-2-methylpyridine