missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)pyridine 98.0+%, TCI America™

Catalog No. B47101G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47101G Supplier TCI America Supplier No. B47101G

missing translation for 'validMainframeMsg'

Specifications

CAS 436799-33-6
Melting Point 44°C
Color White-Yellow
Boiling Point 66°C
Molecular Formula C6H3BrF3N
Quantity 1 g
MDL Number MFCD04972700
UN Number 2811
Synonym 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin
InChI Key HEDHNDVPKRVQPN-UHFFFAOYSA-N
SMILES C1=C(C=NC=C1Br)C(F)(F)F
IUPAC Name 3-bromo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol) 225.996
PubChem CID 11127991
Formula Weight 226.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine
Show More Show Less