missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitropyridine N-oxide, 98+%, Thermo Scientific Chemicals

Numéro de catalogue. AAL1952406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAL1952406 5 g
AAL1952414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAL1952406 Fournisseur Thermo Scientific Chemicals Code fournisseur L1952406
Il en reste null

3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-nitropyridine N-oxide is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Sparingly Soluble in water (0.92 g/L at 25°C).

Notes
Store under inert gas. It is sensitive to air. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Identifiants chimiques

CAS 1678-49-5
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
MDL Number MFCD00085925
InChI Key YZQMKADIENBVLH-UHFFFAOYSA-N
Synonym 3-bromo-4-nitropyridine n-oxide, 3-bromo-4-nitropyridine 1-oxide, 3-bromo-4-nitropyridin-1-ium-1-olate, pyridine, 3-bromo-4-nitro-, 1-oxide, 3-bromo-4-nitropyridine-n-oxide, 3-bromo-4-nitro-1-oxido-pyridin-1-ium, 3-bromo-4-nitropyridin-1-ol, acmc-1bxbw, 3-bromo-4-nitropyridine oxide, 3-bromo-4-nitropyridin-n-oxide
PubChem CID 548521
IUPAC Name 3-bromo-4-nitro-1-oxidopyridin-1-ium
SMILES C1=C[N+](=CC(=C1[N+](=O)[O-])Br)[O-]

Spécifications

Melting Point 152°C to 154°C
Quantity 5 g
Beilstein 150822
Solubility Information Sparingly Soluble in water (0.92g/L at 25°C).
Formula Weight 219
Percent Purity ≥98%
Chemical Name or Material 3-Bromo-4-nitropyridine N-oxide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only