missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methylphenol, 98%

Catalog No. AAH2868906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2868906 5 g
AAH2868903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2868906 Supplier Thermo Scientific Chemicals Supplier No. H2868906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60710-39-6 | C7H7BrO | 187.036 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 60710-39-6
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD08273793
InChI Key GMZKNRDHSHYMHG-UHFFFAOYSA-N
Synonym 3-bromo-p-cresol, 2-bromo-4-hydroxytoluene, phenol, 3-bromo-4-methyl, 3-bromo-4-methyl-phenol, 3-brom-p-kresol, acmc-209mlm, 3-bromo-4-methyl phenol, phenol,3-bromo-4-methyl, 3-bromanyl-4-methyl-phenol, ksc494c1p
PubChem CID 10307913
IUPAC Name 3-bromo-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)Br

Specifications

Melting Point 54°C to 57°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 3-Bromo-4-methylphenol
Hazard Category H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only