missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzonitrile 98.0+%, TCI America™

Catalog No. B31245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B31245G Supplier TCI America Supplier No. B31245G
Only null left

Chemical Identifiers

CAS 117572-79-9
Molecular Formula C8H6BrNO
Molecular Weight (g/mol) 212.05
MDL Number MFCD00143097
InChI Key QHWZMDRKTYTPEE-UHFFFAOYSA-N
Synonym 2-bromo-4-cyanoanisole, 3-bromo-4-methoxylbenzonitrile, 3-bromo-4-methoxybenzenecarbonitrile, 3-bromo-4-methoxy-benzonitrile, benzonitrile, 3-bromo-4-methoxy, pubchem3795, acmc-1cexj, maybridge4_001971, 3-bromo4-methoxybenzonitrile, ksc494m8l
PubChem CID 736049
IUPAC Name 3-bromo-4-methoxybenzonitrile
SMILES COC1=CC=C(C=C1Br)C#N

Specifications

Melting Point 123°C
Quantity 5 g
UN Number 3439
Formula Weight 212.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxybenzonitrile