missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. B33365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33365G 5 g
B333625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33365G Supplier TCI America Supplier No. B33365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 99-58-1
Molecular Formula C8H6BrO3
Molecular Weight (g/mol) 230.04
MDL Number MFCD00020295
InChI Key BBPZABXVRBFWGD-UHFFFAOYSA-M
Synonym 3-bromo-p-anisic acid, benzoic acid, 3-bromo-4-methoxy, 3-bromo-4-methoxybenzoicacid, akos bbb/210, rarechem al bo 0751, timtec-bb sbb009955, 4-methoxy-3-bromo benzoic acid, 3-bromanissaure, pubchem2675, 2-bromo-4-carboxyanisole
PubChem CID 66836
IUPAC Name 3-bromo-4-methoxybenzoate
SMILES COC1=CC=C(C=C1Br)C([O-])=O

Specifications

Melting Point 222°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-methoxybenzoic Acid