missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-chloroaniline 98.0+%, TCI America™

Catalog No. B41601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B41601G Supplier TCI America Supplier No. B41601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 823-54-1
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.47
MDL Number MFCD00173905
InChI Key FVZODFVCIDBDGS-UHFFFAOYSA-N
Synonym benzenamine, 3-bromo-4-chloro, 3-bromo-4-chloro-phenylamine, pubchem5307, acmc-209pod, 3-bromo-4-chlorobenzenamine, ksc493g6f, 3-bromanyl-4-chloranyl-aniline
PubChem CID 12843722
IUPAC Name 3-bromo-4-chloroaniline
SMILES NC1=CC(Br)=C(Cl)C=C1

Specifications

Melting Point 82°C
Color Yellow
Quantity 1 g
Formula Weight 206.47
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-4-chloroaniline