missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-naphthol 96.0+%, TCI America™

Catalog No. B31271G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31271G 1 g
B31275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31271G Supplier TCI America Supplier No. B31271G

Chemical Identifiers

CAS 30478-88-7
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.07
MDL Number MFCD10000951
InChI Key PQVIOPAWVAOHOA-UHFFFAOYSA-N
PubChem CID 45943
IUPAC Name 3-bromonaphthalen-2-ol
SMILES OC1=CC2=CC=CC=C2C=C1Br

Specifications

Melting Point 82°C
Color White-Yellow
Quantity 1 g
Formula Weight 223.07
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-naphthol