missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-hydroxy-5-nitropyridine 97.0+%, TCI America™

Catalog No. p-7123683
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46725G 5 g
B467225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46725G Supplier TCI America Supplier No. B46725G
Only null left

Chemical Identifiers

CAS 15862-33-6
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
MDL Number MFCD03840431
InChI Key KWSOHRDMTWDAOI-UHFFFAOYSA-N
Synonym 3-bromo-2-hydroxy-5-nitropyridine, 3-bromo-5-nitropyridin-2-ol, 2-hydroxy-3-bromo-5-nitropyridine, 3-bromo-5-nitro-2 1h-pyridinone, 3-bromo-5-nitro-2-pyridinol, 2 1h-pyridinone, 3-bromo-5-nitro, 3-bromo-5-nitro-2-hydroxypyridine, pubchem6708, acmc-209dic, ksc684c2p
PubChem CID 613279
IUPAC Name 3-bromo-5-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1[N+](=O)[O-])Br

Specifications

Melting Point 216°C
Color Yellow
Quantity 5 g
Formula Weight 218.99
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Bromo-2-hydroxy-5-nitropyridine