missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH5023506
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5023506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5023506 Supplier Thermo Scientific Chemicals Supplier No. H5023506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 161957-56-8
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00665763
InChI Key UVKURTLVTLRSSM-UHFFFAOYSA-N
Synonym 3-bromo-2-fluoro-benzoic acid, 3-bromo-2-fluorobenzoicacid, 2-fluoro-3-bromobenzoic acid, benzoic acid, 3-bromo-2-fluoro, 3-bromo-2-fluoro benzoic acid, buttpark 14\01-30, 3-bromo-2-fluorobenzenecarboxylic acid, pubchem1318, acmc-209dnk, intermediates-zcf02220
PubChem CID 2736313
IUPAC Name 3-bromo-2-fluorobenzoic acid
SMILES C1=CC(=C(C(=C1)Br)F)C(=O)O

Specifications

Melting Point 168°C to 170°C
Quantity 5 g
Formula Weight 219.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only