missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-fluoro-5-nitropyridine, 97%

Catalog No. AAH6150014
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6150014 25 g
AAH6150003 1 g
AAH6150006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6150014 Supplier Thermo Scientific Chemicals Supplier No. H6150014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1868-58-2 | C5H2BrFN2O2 | 220.99 g/mol

3-Bromo-2-fluoro-5-nitropyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-fluoro-5-nitropyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1868-58-2
Molecular Formula C5H2BrFN2O2
Molecular Weight (g/mol) 220.99
MDL Number MFCD08703668
InChI Key NLLPMLSUMJHTCJ-UHFFFAOYSA-N
Synonym 2-fluoro-3-bromo-5-nitropyridine, acmc-1celf, 3-bromanyl-2-fluoranyl-5-nitro-pyridine
PubChem CID 345124
IUPAC Name 3-bromo-2-fluoro-5-nitropyridine
SMILES [O-][N+](=O)C1=CN=C(F)C(Br)=C1

Specifications

Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 220.98
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-fluoro-5-nitropyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only