missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-ethoxypyridine, 95%, Thermo Scientific Chemicals

Catalog No. AAH2769003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2769003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2769003 Supplier Thermo Scientific Chemicals Supplier No. H2769003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-ethoxypyridin is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 57883-25-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD00234309
InChI Key KLQBFCAXXZMOLZ-UHFFFAOYSA-N
PubChem CID 10987308
IUPAC Name 3-bromo-2-ethoxypyridine
SMILES CCOC1=C(C=CC=N1)Br

Specifications

Quantity 1 g
Synonym 2-ethoxy-3-bromopyridine, pyridine, 3-bromo-2-ethoxy, 3-bromanyl-2-ethoxy-pyridine, 3-bromo-2-ethoxy-pyridine, pubchem6233, bromo-2 ethoxypyridine, 3-bromo-2-ethoxypyridine, pyridine,3-bromo-2-ethoxy
Formula Weight 202.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-2-ethoxypyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only