missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-chlorothiophene 97.0+%, TCI America™

Catalog No. B20135G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20135G 5 g
B201325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20135G Supplier TCI America Supplier No. B20135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 40032-73-3
Molecular Formula C4H2BrClS
Molecular Weight (g/mol) 197.474
MDL Number MFCD00043883
InChI Key KSHOQKKCPJELBV-UHFFFAOYSA-N
Synonym 2-chloro-3-bromo thiophene, 2-chloro-3-bromothiophene, 3-bromo-2-chloro-thiophene, thiophene, 3-bromo-2-chloro, pubchem5259, acmc-1apw4, ksc491i4t, bidd:gt0537, 3-bromo-2-chlorothiophene, inchi=1/c4h2brcls/c5-3-1-2-7-4 3 6/h1-2
PubChem CID 2724559
IUPAC Name 3-bromo-2-chlorothiophene
SMILES C1=CSC(=C1Br)Cl

Specifications

Boiling Point 73°C
Quantity 5 g
Formula Weight 197.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-chlorothiophene