missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-butanone, 95%, stabilized

Catalog No. AC402910050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402910050 5 g Glass bottle
AC402910010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC402910050 Supplier Thermo Scientific Chemicals Supplier No. 402910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 814-75-5 | C4H7BrO | 151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 814-75-5
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151
MDL Number MFCD00013538
InChI Key BNBOUFHCTIFWHN-UHFFFAOYSA-N
Synonym 3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
PubChem CID 13142
IUPAC Name 3-bromobutan-2-one
SMILES CC(C(=O)C)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.4380g/mL
Boiling Point 36.0°C (11.0 mmHg)
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4580 to 1.4600
Linear Formula CH3COCHBrCH3
Quantity 5 g
Specific Gravity 1.438
Stabilizer 1% max. magnesium oxide
Formula Weight 151
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-butanone, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to f
EINECSNumber 212-404-6
RTECSNumber EL7005000
TSCA TSCA

RUO – Research Use Only