missing translation for 'onlineSavingsMsg'
Learn More

3-bromo-1-methyl-1h-pyrazolo[3,4-b]pyridine, 95%, Thermo Scientific™

Catalog No. p-3880568
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC70910CB 250mg Amber Glass Bottle
CC70910DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC70910CB Supplier Thermo Scientific Chemicals Supplier No. CC70910CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 116855-03-9
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD11878034
InChI Key BVDWIHGOGVWCCJ-UHFFFAOYSA-N
Synonym 3-bromo-1-methyl-1h-pyrazolo 3,4-b pyridine, 3-bromo-1-methylpyrazolo 3,4-b pyridine, 3-bromo-1-methylpyrazolo 5,4-b pyridine, 1h-pyrazolo 3,4-b pyridine, 3-bromo-1-methyl
PubChem CID 278149
IUPAC Name 3-bromo-1-methylpyrazolo[3,4-b]pyridine
SMILES CN1C2=C(C=CC=N2)C(=N1)Br

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 212.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-1-methyl-1H-pyrazolo[3, 4-b]pyridine
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.