missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)cyclohexanone, 95%, Thermo Scientific Chemicals

Catalog No. AAH66594MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66594MD 250 mg
AAH6659403 1 g
AAH6659406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66594MD Supplier Thermo Scientific Chemicals Supplier No. H66594MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 885280-38-6
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.28
MDL Number MFCD03790905
InChI Key VGDCXKATFLOEHF-UHFFFAOYNA-N
Synonym 3-n-boc-aminocyclohexanone, tert-butyl 3-oxocyclohexylcarbamate, 3-oxo-cyclohexyl-carbamic acid tert-butyl ester, tert-butyl 3-oxocyclohexyl carbamate, 3-boc-amino cyclohexanone, tert-butyl n-3-oxocyclohexyl carbamate, tert-butyl3-oxocyclohexylcarbamate, pubchem20066, 3-n-boc-amino cyclohexanone, 3-boc-amino cyclohexan-1-one
PubChem CID 44228975
IUPAC Name tert-butyl N-(3-oxocyclohexyl)carbamate
SMILES CC(C)(C)OC(=O)NC1CCCC(=O)C1

Specifications

Beilstein 11040732
Quantity 250 mg
Formula Weight 213.28
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-(Boc-amino)cyclohexanone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only