missing translation for 'onlineSavingsMsg'
Learn More

3-Benzylrhodanine 98.0+%, TCI America™

Catalog No. B165625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B165625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B165625G Supplier TCI America Supplier No. B165625G
Only null left

Chemical Identifiers

CAS 10574-69-3
Molecular Formula C10H9NOS2
Molecular Weight (g/mol) 223.308
MDL Number MFCD00086924
InChI Key ZFHVUMCTGGAWBM-UHFFFAOYSA-N
Synonym 3-Benzyl-2-thioxo-4-thiazolidinone
PubChem CID 97673
IUPAC Name 3-benzyl-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES C1C(=O)N(C(=S)S1)CC2=CC=CC=C2

Specifications

Melting Point 89°C
Quantity 25 g
Formula Weight 223.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzylrhodanine
EINECSNumber (5)-3403
RTECSNumber VI7800000
TSCA No