missing translation for 'onlineSavingsMsg'
Learn More

3-[(Benzyloxycarbonyl)amino]propionaldehyde, 95%

Catalog No. AC427720010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427720010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC427720010 Supplier Thermo Scientific Chemicals Supplier No. 427720010
Only null left

CAS: 65564-05-8 | C11H13NO3 | 207.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 65564-05-8
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.23
MDL Number MFCD05664298
InChI Key PQMOZOQTXKMYSK-UHFFFAOYSA-N
Synonym benzyl 3-oxopropylcarbamate, 3-benzyloxycarbonyl amino-1-propanal, 3-benzyloxycarbonyl amino propionaldehyde, benzyl n-3-oxopropyl carbamate, benzyl 3-oxopropyl carbamate, 3-oxo-propyl-carbamic acid benzyl ester, n-phenylmethoxycarbonyl-3-aminopropanal, n-cbz-3-aminopropanal, pubchem13633
PubChem CID 10398106
IUPAC Name benzyl N-(3-oxopropyl)carbamate
SMILES C1=CC=C(C=C1)COC(=O)NCCC=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 52°C to 57°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2OC(O)NHCH2CH2CHO
Quantity 1 g
Formula Weight 207.23
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-[(Benzyloxycarbonyl)amino]propionaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only