missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxybenzyl Alcohol 97.0+%, TCI America™

Catalog No. B38575G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38575G 5 g
B385725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38575G Supplier TCI America Supplier No. B38575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1700-30-7
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
MDL Number MFCD00004642
InChI Key AFKLSWIRJUJWKY-UHFFFAOYSA-N
Synonym 3-benzyloxybenzyl alcohol, 3-benzyloxy phenyl methanol, 3-benzyloxybenzylalcohol, benzenemethanol, 3-phenylmethoxy, 3-phenylmethoxyphenyl methanol, 3-benzyloxyphenyl methanol, 3-phenylmethoxy phenyl methan-1-ol, pubchem7731, acmc-1bo1a
PubChem CID 74341
IUPAC Name (3-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC(=C2)CO

Specifications

Melting Point 49°C
Boiling Point 160°C
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Benzyloxybenzyl Alcohol