missing translation for 'onlineSavingsMsg'
Learn More

3'-Benzyloxyacetophenone 98.0+%, TCI America™

Catalog No. B47055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47055G 5 g
B470525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47055G Supplier TCI America Supplier No. B47055G

Chemical Identifiers

CAS 34068-01-4
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.275
MDL Number MFCD00026221
InChI Key FGQMEAWGAUALJQ-UHFFFAOYSA-N
Synonym 1-3-benzyloxy phenyl ethanone, 3-benzyloxyacetophenone, 3'-benzyloxyacetophenone, 1-3-benzyloxy phenyl ethan-1-one, 1-3-phenylmethoxy phenyl ethan-1-one, 3'-benzyloxy acetophenone, ethanone, 1-3-phenylmethoxy phenyl, 1-acetyl-3-phenylmethoxy benzene
PubChem CID 98689
IUPAC Name 1-(3-phenylmethoxyphenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)OCC2=CC=CC=C2

Specifications

Color White-Yellow
Boiling Point 154°C
Quantity 5 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3′-Benzyloxyacetophenone