missing translation for 'onlineSavingsMsg'
Learn More

3-Aminosalicylic Acid 98.0+%, TCI America™

Catalog No. A04215G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A04215G 5 g
A04211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A04215G Supplier TCI America Supplier No. A04215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 570-23-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00010299
InChI Key IQGMRVWUTCYCST-UHFFFAOYSA-N
Synonym 3-aminosalicylic acid, 3-amino salicylic acid, benzoic acid, 3-amino-2-hydroxy, 3-amino-2-hydroxy-benzoic acid, 2-amino-6-carboxyphenol, salicylic acid, 3-amino, 3-carboxy-2-hydroxyaniline, 3 or 5-aminosalicylic acid, benzoic acid, amino-2-hydroxy
PubChem CID 68443
IUPAC Name 3-amino-2-hydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)N)O)C(=O)O

Specifications

Melting Point 221°C
Quantity 5 g
Formula Weight 153.14
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminosalicylic Acid