missing translation for 'onlineSavingsMsg'
Learn More

3-Aminoquinuclidine Dihydrochloride 98.0+%, TCI America™

Catalog No. A12365G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12365G 5 g
A123625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12365G Supplier TCI America Supplier No. A12365G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6530-09-2
Molecular Formula C7H16Cl2N2
Molecular Weight (g/mol) 199.12
MDL Number MFCD00137395
InChI Key STZHBULOYDCZET-UHFFFAOYNA-N
Synonym 3-aminoquinuclidine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, 1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, quinuclidine-3-ylamine, chloride, chloride
PubChem CID 197853
IUPAC Name 1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride
SMILES C1CN2CCC1C(C2)N.Cl.Cl

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 199.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminoquinuclidine Dihydrochloride
RTECSNumber VD4590000
TSCA No