missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminopyridine, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 60.0°C to 63.0°C |
| Color | Colorless to Yellow |
| Boiling Point | 248.0°C |
| Flash Point | 124°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (C5H4N)NH2 |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Danger |
| missing translation for 'healthHazard2' | GHS H Statement Fatal if swallowed. Toxic in contact with skin. May cause respiratory irritation. Causes serious eye irritation. Causes skin irritation. |
| missing translation for 'healthHazard3' | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Gently wash with plenty of soap and water. IF IN EYES: Rinse cautiously wit |
| EINECSNumber | 207-322-2 |
| RTECSNumber | US1650000 |
| TSCA | TSCA |
RUO – Research Use Only