missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. A08485G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A08485G 5 g
A084825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A08485G Supplier TCI America Supplier No. A08485G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1-1-5424
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006141
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
Synonym 3-amino-2-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 3-amino, 2-amino-3-pyrazinecarboxylic acid, 3-aminopiperazine-2-carboxylic acid, 2-aminopyrazine-3-carboxylic acid, 2-pyrazinecarboxylic acid, 3-amino, 3-aminopyrazine-2-carboxyliacid, zlchem 459, pubchem8544, 3-aminopyrazinoic acid
PubChem CID 72656
IUPAC Name 3-aminopyrazine-2-carboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)N

Specifications

Melting Point 195°C
Color Yellow
Quantity 5 g
Formula Weight 139.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyrazine-2-carboxylic Acid