missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopyrazine-2-carboxylic acid, 99+%

Catalog No. AC104500250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104500250 25 g Glass bottle
AC104500050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104500250 Supplier Thermo Scientific Chemicals Supplier No. 104500250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5424-01-1 | C5H5N3O2 | 139.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1-1-5424
Molecular Formula C5H5N3O2
Molecular Weight (g/mol) 139.11
MDL Number MFCD00006141
InChI Key ZAGZIOYVEIDDJA-UHFFFAOYSA-N
Synonym 3-amino-2-pyrazinecarboxylic acid, pyrazinecarboxylic acid, 3-amino, 2-amino-3-pyrazinecarboxylic acid, 3-aminopiperazine-2-carboxylic acid, 2-aminopyrazine-3-carboxylic acid, 2-pyrazinecarboxylic acid, 3-amino, 3-aminopyrazine-2-carboxyliacid, zlchem 459, pubchem8544, 3-aminopyrazinoic acid
PubChem CID 72656
IUPAC Name 3-aminopyrazine-2-carboxylic acid
SMILES C1=CN=C(C(=N1)C(=O)O)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 201°C to 205°C
Color Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Formula Weight 139.11
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopyrazine-2-carboxylic acid, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 226-558-7
TSCA TSCA

RUO – Research Use Only