missing translation for 'onlineSavingsMsg'
Learn More

3-Aminopropyltrimethoxysilane, 95%

Catalog No. AC313255000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC313255000 500 mL Glass bottle
AC313251000 100 mL Glass bottle
AC313250025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC313255000 Supplier Thermo Scientific Chemicals Supplier No. 313255000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13822-56-5 | C6H17NO3Si | 179.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.29
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 1.0100g/mL
Boiling Point 194.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.4230 to 1.4250
Linear Formula NH2(CH2)3Si(OCH3)3
Quantity 500 mL
Specific Gravity 1.01
Solubility Information Solubility in water: reacts with water.
Formula Weight 179.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Aminopropyltrimethoxysilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
EINECSNumber 237-511-5

RUO – Research Use Only