missing translation for 'onlineSavingsMsg'
Learn More

3-Aminophthalic acid hydrochloride dihydrate, 96%

Catalog No. AAA1031914
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1031914 25 g
AAA1031903 1 g
AAA1031906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1031914 Supplier Thermo Scientific Chemicals Supplier No. A1031914
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1852533-96-0 | C8H8ClNO4 | 217.605 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1852533-96-0
Molecular Formula C8H8ClNO4
Molecular Weight (g/mol) 217.605
MDL Number MFCD00150282
InChI Key ZBZAVEORKXFUQB-UHFFFAOYSA-N
Synonym 3-aminophthalic acid hydrochloride, 1,2-benzenedicarboxylic acid, 3-amino-, hydrochloride, 1,2-benzenedicarboxylic acid, 3-amino-, hydrochloride 1:1, phthalic acid, hydrochloride, 1, 3-amino-, hydrochloride, 3-aminobenzene-1,2-dicarboxylic acid hydrochloride, 3-amino phthalic acid hydrochloride, phthalic acid, 3-amino-, hydrochloride, acmc-1b7k3, ghl.pd_mitscher_leg0.1315
PubChem CID 81375
IUPAC Name 3-aminophthalic acid;hydrochloride
SMILES C1=CC(=C(C(=C1)N)C(=O)O)C(=O)O.Cl

Specifications

Melting Point 182°C (decomposition)
Quantity 25 g
Formula Weight 253.65 (217.62 Anhydrous)
Percent Purity 98%
Chemical Name or Material 3-Aminophthalic acid hydrochloride dihydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 230-106-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only