missing translation for 'onlineSavingsMsg'
Learn More

3-Aminobenzotrifluoride 99.0+%, TCI America™

Catalog No. A0434500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0434500G 500 g
A043425G 25 g
A0434100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0434500G Supplier TCI America Supplier No. A0434500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-16-8
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007797
InChI Key VIUDTWATMPPKEL-UHFFFAOYSA-N
Synonym 3-trifluoromethyl aniline, 3-aminobenzotrifluoride, m-aminobenzotrifluoride, 3-aminotrifluorotoluene, benzenamine, 3-trifluoromethyl, 3-trifluoromethyl benzenamine, m-abtf, m-trifluoromethylaniline, m-amino trifluorotoluene, m-trifluoromethyl aniline
PubChem CID 7375
IUPAC Name 3-(trifluoromethyl)aniline
SMILES C1=CC(=CC(=C1)N)C(F)(F)F

Specifications

Melting Point 3°C
Color Yellow
Boiling Point 189°C
Quantity 500 g
UN Number 2948
Formula Weight 161.13
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Aminobenzotrifluoride
EINECSNumber (3)-0223
RTECSNumber XU9180000
TSCA Yes