missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Aminobenzotrifluoride, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| CAS Min % | 98 |
| Melting Point | 5.0°C to 6.0°C |
| Color | Yellow |
| Density | 1.3030g/mL |
| Boiling Point | 187.0°C to 193.0°C |
| Flash Point | 85°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.4790 to 1.4810 |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Harmful in contact with skin. Fatal if inhaled. Causes skin irritation. Causes serious eye damage. May cause respiratory irritation. May cause damage to organs through prolo |
| Precautionary Statement | GHS P Statement Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN: Wash w |
| EINECSNumber | 202-643-4 |
| RTECSNumber | XU9180000 |
| TSCA | TSCA |
| Recommended Storage | This product may darken on storage |
RUO – Research Use Only