missing translation for 'onlineSavingsMsg'
Learn More

Acide 3-aminobenzénéboronique hémisulfate, 98+%

Numéro de catalogue. AAA1724003
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1724003 1 g
AAA1724006 5 g
AAA1724014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. AAA1724003 missing translation for 'mfr' Thermo Scientific Chemicals missing translation for 'supplierNo' A1724003

CAS : 66472-86-4 | C12H18B2N2O8S | 371,96 g/mol

Ce Thermo Scientific Chemicals produit de marque faisait à l’origine partie du portefeuille de Alfa Aesar produits. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit / article original Alfa Aesar ou la référence au SKU n’a pas changé dans le cadre de la transition de la marque vers Thermo Scientific Chemicals.

Chemical Identifiers

CAS 66472-86-4
Molecular Formula C12H18B2N2O8S
Molecular Weight (g/mol) 371.96
MDL Number MFCD00013111
InChI Key UKTAURVTSWDIQR-UHFFFAOYSA-N
Synonym 3-aminophenyl boronic acid sulfate 2:1, 3-aminobenzeneboronic acid hemisulfate salt, 3-aminophenylboronic acid hemisulfate, 3-aminobenzeneboronic acid hemisulfate, 3-aminophenylboronic acid hemisulphate, 4-aminophenylboronic acid hemisulfate, m-aminophenyl boronic acid, hemisulphate, boronic acid, 3-aminophenyl-, sulfate 2:1, 3-aminophenyl boronic acid; sulfuric acid, bis m-aminophenylboronic acid ; sulfuric acid
PubChem CID 16211139
IUPAC Name acide boronique (3-aminophényl); Acide sulfurique
SMILES OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 8094151
Sensitivity Hygroscopic
Formula Weight 185.98
Percent Purity ≥98%
Chemical Name or Material 3-Aminobenzeneboronic acid hemisulfate
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 266-376-5
TSCA No
Recommended Storage Température ambiante

RUO – Usage réservé à la recherche