missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-(trifluoromethyl)benzoic Acid 97.0+%, TCI America™

Catalog No. A24811G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24811G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24811G Supplier TCI America Supplier No. A24811G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 328-68-7
Molecular Formula C8H6F3NO2
Molecular Weight (g/mol) 205.14
MDL Number MFCD00236641
InChI Key WBTHOSZMTIPJLR-UHFFFAOYSA-N
Synonym 3-amino-5-trifluoromethyl benzoic acid, 3-amino-5-trifluoromethyl benzoicacid, 5-amino-3-trifluoromethyl benzoic acid, benzoic acid, 3-amino-5-trifluoromethyl, pubchem1359, acmc-209xwk, ksc495q1r, 3-amino-5-carboxybenzotrifluoride, buttpark 25\01-23, 3-carboxy-5-trifluoromethyl aniline
PubChem CID 609556
IUPAC Name 3-amino-5-(trifluoromethyl)benzoic acid
SMILES NC1=CC(=CC(=C1)C(F)(F)F)C(O)=O

Specifications

Melting Point 143°C
Quantity 1 g
Formula Weight 205.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-(trifluoromethyl)benzoic Acid