missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-fluoropyridine 97,0+%, TCI America™

Numéro de catalogue. A27531G
Modifier l'affichage
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A27531G 1 g
A27535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. A27531G Fournisseur TCI America Code fournisseur A27531G

Identifiants chimiques

CAS 210169-05-4
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.107
MDL Number MFCD00234073
InChI Key ZRORIJXOWXYPMO-UHFFFAOYSA-N
Synonym 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine
PubChem CID 574450
IUPAC Name 5-fluoropyridine-3-amine
SMILES C1=C(C=NC=C1F)N

Spécifications

Melting Point 87°C
Color Jaune
Quantity 1 g
Formula Weight 112.11
Percent Purity ≥97.0% (GC,T)
Physical Form Poudre cristalline
Chemical Name or Material 3-Amino-5-fluoropyridine