missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-1H-indazole, 97%, Thermo Scientific Chemicals

Catalog No. AAH3335103
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3335103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3335103 Supplier Thermo Scientific Chemicals Supplier No. H3335103
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61272-71-7
Molecular Formula C7H6BrN3
Molecular Weight (g/mol) 212.05
MDL Number MFCD03426696
InChI Key OMPYFDJVSAMSMA-UHFFFAOYSA-N
Synonym 3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine
PubChem CID 817910
IUPAC Name 5-bromo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=C(Br)C=C12

Specifications

Melting Point 171°C to 174°C
Quantity 1 g
Formula Weight 212.05
Percent Purity 97%
Chemical Name or Material 3-Amino-5-bromo-1H-indazole
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only