missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-1-methyl-1H-indazole, 97%, Thermo Scientific™

Catalog No. AAH3320203
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3320203 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3320203 Supplier Thermo Scientific Chemicals Supplier No. H3320203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

CAS 1000018-06-3
Molecular Formula C8H8BrN3
Quantity 1g
MDL Number MFCD09864804
Synonym 5-bromo-1-methyl-1h-indazol-3-amine, 3-amino-5-bromo-1-methyl-1h-indazole, acmc-2097ln, 5-bromo-1-methyl-1h-indazole-3-ylamine
InChI Key BKBSBRJIGMVBFM-UHFFFAOYSA-N
SMILES CN1C2=C(C=C(C=C2)Br)C(=N1)N
IUPAC Name 5-bromo-1-methylindazol-3-amine
Molecular Weight (g/mol) 226.077
PubChem CID 26369215
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 3-Amino-5-bromo-1-methyl-1H-indazole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only